Anti-S100 A8 Mouse Monoclonal Antibody [clone: MAC387]
Catalog # 89362-586
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:S100 Calcium binding Protein A8
- Antigen symbol:S100 A8
- Clonality:Monoclonal
- Clone:MAC387
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Baboon
- Size:100 μg
- Antigen synonyms:L1Ag|CP-10|CFAG|CAGA|MRP8|NIF|MIF|CGLA|60B8AG|MA387|P8
- Gene ID:6280
- Immunogen:Human monocytes
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Storage buffer:Phosphate buffered saline containing 0.09% Sodium Azide 1% Bovine Serum Albumin
- Storage temperature:–20 °C
- Tested applications:Fluorescence-Activated Cell Sorting
- Supplier No.:GTX27429
Specifications
About this item
Mouse monoclonal antibody [MAC387] to Macrophage + Monocyte
Type: Primary
Antigen: S100 A8
Clonality: Monoclonal
Clone: MAC387
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Baboon
Frequently Bought Together