Order Entry
ContactUsLinkComponent
Anti-RBL2 Mouse Monoclonal Antibody [clone: KAB40]
Anti-RBL2 Mouse Monoclonal Antibody [clone: KAB40]
Catalog #: 89354-380
Supplier:  Genetex
Anti-RBL2 Mouse Monoclonal Antibody [clone: KAB40]
Catalog #: 89354-380
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RB Transcriptional Corepressor Like 2
  • Clonality:
    Monoclonal
  • Clone:
    KAB40
  • Gene ID:
    5934
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Donkey
  • Antigen symbol:
    RBL2
  • Conjugation:
    Unconjugated
  • Size:
    50 μg
  • Western blot:
    Yes
  • Antigen synonyms:
    RBR 2,
    RBL2,
    FLJ26459,
    Retinoblastoma like 2,
    130 kDa retinoblastoma associated protein,
    Retinoblastoma like protein 2,
    Rb2,
    Rb 2,
    PRB 2,
    p130,
    PRB2,
    RBR2,
    Retinoblastoma Related Gene.
  • Storage buffer:
    Phosphate-buffered saline, pH 7.2, containing 0.08% Sodium Azide
  • Storage temperature:
    Upon Receipt - Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1mg/ml
  • Immunogen:
    Recombinant fusion protein (Human).
  • Tested applications:
    WB
  • Purification:
    Protein G purified
  • Cat. no.:
    89354-380
  • Supplier No.:
    GTX10576

Specifications

About this item

Mouse monoclonal [KAB40] to Rb2 p130

WB: Use at a dilution of 1/1000. Detects a band of approximately 130 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: RBL2
Clonality: Monoclonal
Clone: KAB40
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Donkey

Customers Who Bought This Also Bought