Order Entry
ContactUsLinkComponent
Anti-P2RY1 Rabbit Polyclonal Antibody
Anti-P2RY1 Rabbit Polyclonal Antibody
Catalog #: 89365-250
Supplier:  Genetex
Anti-P2RY1 Rabbit Polyclonal Antibody
Catalog #: 89365-250
Supplier:  Genetex
Price
$760.80
Log in for availability
Unit of Measure Each (25µG)
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Purinergic Receptor P2Y, G-protein coupled 1
  • Clonality:
    Polyclonal
  • Gene ID:
    5028
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    P2RY1
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Antigen synonyms:
    P2Y,
    P2y1
  • Storage buffer:
    Phosphate buffered saline, PH 7.7 containing 0.01% sodium azide
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive)
  • Tested applications:
    IHC-P
  • Purification:
    Immunogen affinity purified.
  • Cat. no.:
    89365-250

Specifications

About this item

Rabbit Polyclonal to Human Purinergic Receptor P2Y1

Immunohistochemistry with formalin fixed paraffin embedded tissues. Not tested for other uses. IHC-P: Use at a dilution of 1/100 - 1/200.

Type: Primary
Antigen: P2RY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Customers Who Bought This Also Bought