Order Entry
ContactUsLinkComponent
Anti-PGS1 Rabbit Polyclonal Antibody
Anti-PGS1 Rabbit Polyclonal Antibody
Catalog #: 89363-598
Supplier:  Genetex
Anti-PGS1 Rabbit Polyclonal Antibody
Catalog #: 89363-598
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Phosphatidylglycerophosphate Synthase 1
  • Clonality:
    Polyclonal
  • Gene ID:
    9489
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    PGS1
  • Conjugation:
    Unconjugated
  • Size:
    100 μg
  • Western blot:
    Yes
  • Storage buffer:
    Phosphate buffered saline (PBS)
  • Molecular weight:
    60 kDa
  • Storage temperature:
    Store at 4 °C for short-term use. For longer periods of storage, aliquot and store at –20 °C. Avoid repeat freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Synthetic peptide
  • Tested applications:
    WB
  • Purification:
    Protein A Purified Total IgG
  • Cat. no.:
    89363-598
  • Supplier No.:
    GTX46172

Specifications

About this item

Rabbit polyclonal antibody to PGS1 (C-terminal)

Recommended Dilutions: For Western Blot: Use at 1.25 ug/ml.

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: PGS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Customers Who Bought This Also Bought