Anti-PAX7 Rabbit Polyclonal Antibody
Catalog # 89348-632
Supplier: Avantor
Specifications
- Antibody type:Primary
- Antigen name:Paired Box 7
- Antigen symbol:PAX7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:5081
- Antigen synonyms:PAX7 transcriptional factor|PAX7A|paired domain gene 7|RMS2|Hup1|HUP1|paired box gene 7|PAX7B|paired box protein Pax-7|Paired box homeotic gene 7
- Storage buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular weight:57 kDa
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PAX7. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89348-632
- Supplier No.:GTX100702
Specifications
About this item
Rabbit Polyclonal antibody to PAX7 (paired box 7)
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: PAX7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human