Order Entry
ContactUsLinkComponent
Anti-P2Y11 Rabbit Polyclonal Antibody
Anti-P2Y11 Rabbit Polyclonal Antibody
Catalog #: 89359-590
Supplier:  Genetex
Anti-P2Y11 Rabbit Polyclonal Antibody
Catalog #: 89359-590
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Purinergic Receptor P2Y G protein coupled 11
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    P2Y11
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Antigen synonyms:
    P2Y11 receptor,
    Purinergic receptor P2Y11.,
    Purinergic receptor P2Y G protein coupled 11,
    P2Y purinoceptor 11,
    P2Y11_HUMAN,
    P2Y11
  • Storage buffer:
    Preservative: 0.01% Sodium Azide; Constituents: PBS. pH 7.7
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive)
  • Tested applications:
    IHC-P
  • Purification:
    Immunogen affinity purified
  • Cat. no.:
    89359-590
  • Supplier No.:
    GTX13104

Specifications

About this item

P2Y11 belongs to the family of G protein coupled receptors, and is coupled to the stimulation of the phosphoinositide and adenylyl cyclase pathways and behaves as a selective purinoceptor.

Type: Primary
Antigen: P2Y11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Customers Who Bought This Also Bought