Order Entry
ContactUsLinkComponent
Anti-PTGES3 Rabbit Polyclonal Antibody
Anti-PTGES3 Rabbit Polyclonal Antibody
Catalog #: 89354-730
Supplier:  Genetex
Price
$920.76
Log in for availability
Unit of Measure Each (100µl)
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Prostaglandin E Synthase 3
  • Clonality:
    Polyclonal
  • Gene ID:
    10728
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    PTGES3
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Antigen synonyms:
    Co chaperone p23
    Progesterone receptor complex
    Sid 3177
    Telomerase binding protein p23
    TEBP
    Hsp90 co chaperone
  • Storage buffer:
    1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Molecular weight:
    19 kDa
  • Storage temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human p23. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89354-730
  • Size:
    100 μl

Specifications

About this item

Rabbit Polyclonal antibody to p23 (prostaglandin E synthase 3 (cytosolic))

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PTGES3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human