Anti-STK24 Rabbit Polyclonal Antibody
89354-980
: Genetex
- Antibody type:Primary
- Antigen name:Serine/Threonine Kinase 24
- Antigen symbol:STK24
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:8428
- Antigen synonyms:STE20|C76483|MST-3|Mst3
- Storage buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular weight:48 kDa
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.51mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human STK24. The exact sequence is proprietary.
- Tested applications:ICC/IF, WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89354-980
- Supplier No.:GTX107500
Rabbit Polyclonal antibody to STK24 (serine/threonine kinase 24 (STE20 homolog, yeast))
Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: STK24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human