Order Entry
ContactUsLinkComponent
Anti-ORAI1 Rabbit Polyclonal Antibody
Anti-ORAI1 Rabbit Polyclonal Antibody
Catalog #: 89366-808
Supplier:  Genetex
Anti-ORAI1 Rabbit Polyclonal Antibody
Catalog #: 89366-808
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ORAI calcium release-activated calcium modulator 1
  • Clonality:
    Polyclonal
  • Gene ID:
    84876
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    ORAI1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • Size:
    100 μg
  • Western blot:
    Yes
  • Antigen synonyms:
    ORAI calcium release-activated calcium modulator 1,
    ORAI1,
    FLJ14466,
    CRACM1,
    calcium release-activated calcium channel protein 1,
    ORAT1,
    calcium release-activated calcium modulator 1,
    TMEM142A,
    transmembrane protein 142A
  • Storage buffer:
    Antibody is supplied in PBS containing 0.02% sodium azide
  • Storage temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Tested applications:
    ELISA, IHC, WB
  • Purification:
    Affinity chromatography purified via peptide column
  • Cat. no.:
    89366-808
  • Supplier No.:
    GTX85048

Specifications

About this item

Rabbit polyclonal antibody to Orai1

Type: Primary
Antigen: ORAI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Customers Who Bought This Also Bought