To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Optic Atrophy 1
- Clonality:Polyclonal
- Gene ID:4976
- Host:Rabbit
- Isotype:IgG
- Reactivity:Chicken
- Antigen symbol:OPA1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:50 μl
- Western blot:Yes
- Antigen synonyms:optic atrophy protein 1|mitochondrial dynamin-like 120 kDa protein|largeG|NTG|mitochondrial|MGM1|dynamin-like guanosine triphosphatase|dynamin-like 120 kDa protein|NPG|mitochondrial dynamin-like GTPase
- Storage buffer:Tris-glycine, 150mM NaCl
- Storage temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:1.04 mg/ml
- Immunogen:A synthetic peptide made to an internal region within residues 500-600 of the human protein. [Swiss-Prot# O60313]
- Tested applications:WB
- Purification:Immunogen affinity purified
- Cat. no.:89363-670
- Supplier No.:GTX48589
Specifications
About this item
Rabbit Polyclonal antibody to OPA1
immunohistochemistry 2.5ug/ml, Western Blot 2 ug/ml
Type: Primary
Antigen: OPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken