Order Entry
ContactUsLinkComponent
Anti-NDMAR2B Rabbit Polyclonal Antibody
Anti-NDMAR2B Rabbit Polyclonal Antibody
Catalog #: 89362-918
Supplier:  Genetex
Anti-NDMAR2B Rabbit Polyclonal Antibody
Catalog #: 89362-918
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    NMDAR2B (phospho Tyr1472)
  • Clonality:
    Polyclonal
  • Gene ID:
    24410
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Antigen symbol:
    NDMAR2B
  • Conjugation:
    Unconjugated
  • Size:
    100 μl
  • Western blot:
    Yes
  • Modification:
    phospho-Tyr1472
  • Storage buffer:
    100 µl in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 µg per ml BSA and 50% glycerol.
  • Molecular weight:
    180 kDa
  • Storage temperature:
    For long-term storage, aliquot and store at -20 C. Avoid repeated freeze/thaw cycles.
  • Immunogen:
    Synthetic phospho-peptide corresponding to amino acid residues surrounding Tyr1472 conjugated to KLH
  • Tested applications:
    WB
  • Purification:
    Antigen affinity purification
  • Cat. no.:
    89362-918
  • Supplier No.:
    GTX30452

Specifications

About this item

Rabbit polyclonal antibody to NMDA, NR2B Subunit (phospho Tyr1472)

Type: Primary
Antigen: NDMAR2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Customers Who Bought This Also Bought