Anti-NAT2 Rabbit Polyclonal Antibody
Catalog # 89357-852
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:N-Acetyltransferase 2 (Arylamine N-Acetyltransferase)
- Antigen symbol:NAT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:10
- Antigen synonyms:PNAT|AAC2|NAT-2
- Storage buffer:0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular weight:34 kDa
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.45mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human NAT2. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89357-852
- Supplier No.:GTX114051
Specifications
About this item
Rabbit Polyclonal antibody to NAT2 (N-acetyltransferase 2 (arylamine N-acetyltransferase))
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: NAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human