Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin E
- Antigen symbol:IgE
- Clonality:Monoclonal
- Clone:LO-ME-3
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Reactivity:Mouse
- Size:125 μg
- Epitope:heavy chain
- Gene ID:380792
- Storage buffer:PBS, with 0.09% sodium azide and 50.0% BSA as preservative.
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Purified IgE from BALB/c mice
- Tested applications:Fluorescence-Activated Cell Sorting
- Purification:Affinity purified with antigen
- Cat. no.:89358-426
- Supplier No.:GTX11581
Specifications
About this item
IgE is the class of antibodies produced in the lungs, skin, and mucous membranes. It may protect against parasite invasion. But it is a major factor in allergic reactions. The antigen-specific IgE interacts with mast cells and eosinophils, triggers the release of histamine, leukotrienes and other substances that lead to the itching, sneezing and congestion of allergies - and the life threatening respiratory distress of asthma and anaphylactic shock.
Recommended Starting Dilutions:FACS: Use at a concentration of 3 - 7 µg/mL. Use 50 µL of this concentration to label 10^6 cells in 100ul.Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Secondary
Antigen: IgE
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: heavy chain
Host: Rat
Isotype: IgG1
Reactivity: Mouse