Order Entry
ContactUsLinkComponent
Anti-MOG Rabbit Polyclonal Antibody
Anti-MOG Rabbit Polyclonal Antibody
Catalog #: 89354-576
Supplier:  Genetex
Anti-MOG Rabbit Polyclonal Antibody
Catalog #: 89354-576
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Myelin Oligodendrocyte Glycoprotein
  • Clonality:
    Polyclonal
  • Gene ID:
    4340
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Antigen symbol:
    MOG
  • Conjugation:
    Unconjugated
  • Size:
    100 μl
  • Antigen synonyms:
    BTNL11,
    NRCLP7,
    MOGIG2,
    BTN6
  • Storage buffer:
    1XPBS, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Molecular weight:
    25 kDa
  • Storage temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1mg/ml
  • Immunogen:
    Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human MOG. The exact sequence is proprietary.
  • Tested applications:
    ELISA-Ag
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. no.:
    89354-576
  • Supplier No.:
    GTX106283

Specifications

About this item

Rabbit polyclonal antibody to MOG (myelin oligodendrocyte glycoprotein)

  • Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: MOG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Customers Who Bought This Also Bought