Order Entry
Canada
ContactUsLinkComponent
Anti-GPR4 Rabbit Polyclonal Antibody
Anti-GPR4 Rabbit Polyclonal Antibody
  89365-042
 :  Genetex
Anti-GPR4 Rabbit Polyclonal Antibody
  89365-042
 :  Genetex
 :  GTX70541

 

  • Antibody type:
    Primary
  • Antigen name:
    G Protein-coupled Receptor 4
  • Antigen symbol:
    GPR4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    25 μg
  • Gene ID:
    2828
  • Antigen synonyms:
    G-protein coupled receptor 19|G-protein coupled receptor 4|G-protein coupled receptor 4,GPR19
  • Storage buffer:
    Phosphate buffered saline, PH 7.7 containing 0.01% sodium azide
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Synthetic peptide - KLH conjugated
  • Tested applications:
    IHC-P
  • Purification:
    Immunogen affinity purified.
  • Cat. no.:
    89365-042
  • Supplier No.:
    GTX70541

 

 

Rabbit Polyclonal to Human G Protein-Coupled Receptor GPR4

For IHC-P: Use at a concentration of 8-16 ?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: GPR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human