Order Entry
ContactUsLinkComponent
Anti-LXR beta Goat Polyclonal Antibody
Anti-LXR beta Goat Polyclonal Antibody
Catalog #: 89367-002
Supplier:  Genetex
Anti-LXR beta Goat Polyclonal Antibody
Catalog #: 89367-002
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Liver X receptor beta
  • Clonality:
    Polyclonal
  • Gene ID:
    7376
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Dog
  • Antigen symbol:
    LXRB
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • Size:
    100 μg
  • Western blot:
    Yes
  • Antigen synonyms:
    LXRB,
    LXR-β,
    RIP15,
    LXR-b,
    UNR,
    NER-I,
    NER
  • Storage buffer:
    Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
  • Storage temperature:
    Aliquot and store at –20 °C. Minimize freezing and thawing.
  • Concentration:
    0.5mg/ml
  • Immunogen:
    SSPTTSSLDTPLPGC
  • Tested applications:
    ELISA, IHC, WB
  • Purification:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Cat. no.:
    89367-002
  • Supplier No.:
    GTX89661

Specifications

About this item

Goat polyclonal antibody to LXR beta / NR1H2

Experiments gave a band at approx 80kDa and a weak band at appr. 60-kDa in human bone marrow lysate after 1µg/ml antibody staining. These bands correspond to earlier findings with different antibodies (PMID: 12393874-60kDa and other unpublished antibodies-80kDa). This protein has a calculated MW of 51.0kDa according to NP_009052. Recommended concentration: 1-3µg/ml. In paraffin embedded Human Spleen shows nucleolar staining. Recommended concentration, 1-2µg/ml. Based on the peptide used, this antibody is expected to be specific for LXR beta and not to cross-react with LXR alpha.

Type: Primary
Antigen: LXR beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Dog

Customers Who Bought This Also Bought