Order Entry
ContactUsLinkComponent
Anti-LC3B Rabbit Polyclonal Antibody
Anti-LC3B Rabbit Polyclonal Antibody
Catalog #: 89348-308
Supplier:  Genetex
RB PAB TO LC3B
Anti-LC3B Rabbit Polyclonal Antibody
Catalog #: 89348-308
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Microtubule-associated Protein 1 light chain 3 beta
  • Clonality:
    Polyclonal
  • Gene ID:
    64862
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    LC3B
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Size:
    100 μl
  • Western blot:
    Yes
  • Antigen synonyms:
    MAP1LC3B,
    Autophagy-related protein LC3 B,
    MAP1 light chain 3-like protein 2,
    MAP1ALC3,
    MAP1A/MAP1B LC3 B,
    Microtubule-associated proteins 1A/1B light chain 3B [Precursor],
    MAP1A/1B light chain 3 B,
    Autophagy-related ubiquitin-like modifier LC3 B
  • Storage buffer:
    1XPBS, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Molecular weight:
    15 kDa
  • Storage temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Full length rat LC3B Recombinant protein.
  • Tested applications:
    WB
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. no.:
    89348-308
  • Supplier No.:
    GTX100240

Specifications

About this item

Rabbit Polyclonal antibody to LC3B (microtubule-associated protein 1 light chain 3 beta)

Suggested starting dilutions are as follows: WB: 1:500-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: LC3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Customers Who Bought This Also Bought