To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Linker for activation of T-cells Family member 1
- Antigen symbol:LAT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 μl
- Antigen synonyms:36 kDa phospho-tyrosine adapter protein|pp36|p36-38|LAT|Linker for activation of T-cells family member 1
- Gene ID:27040
- Modification:phospho-Tyr132
- Concentration:0.35 mg/ml
- Immunogen:Synthetic phospho peptide (Human) derived from a region of LAT that contains tyrosine 132.
- Purification:Immunogen affinity purified
- Storage buffer:Preservative: 0.05% Sodium Azide; Constituents: PBS, 1mg/ml BSA. pH 7.3
- Storage temperature:Aliquot and store at –20 °C or –80 °C. Avoid repeated freeze / thaw cycles.
- Tested applications:WB
- Cat. no.:89361-452
- Supplier No.:GTX24476
Specifications
About this item
Rabbit polyclonal to LAT ( phospho Y132 )
WB: Use at a concentration of 0.1 - 1.0 ?g/ml. Detects a band of approximately 38 kDa (predicted molecular weight: 31.4 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: LAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human