Order Entry
ContactUsLinkComponent
Anti-Ig kappa Mouse Monoclonal Antibody
Anti-Ig kappa Mouse Monoclonal Antibody
Catalog #: 89353-180
Supplier:  Genetex
Anti-Ig kappa Mouse Monoclonal Antibody
Catalog #: 89353-180
Supplier:  Genetex
Price
$774.67
Log in for availability
Unit of Measure Each (1ml)
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Immunoglobulin kappa
  • Clonality:
    Monoclonal
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Antigen symbol:
    Ig kappa
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Epitope:
    Light Chain
  • Storage buffer:
    Diluted ascites containing sodium azide
  • Storage temperature:
    Keep as concentrated solution, aliquot and store at 4°C.
  • Immunogen:
    Purified Bence Jones kappa light chain protein.
  • Tested applications:
    IHC-P, IHC-Fr
  • Purification:
    Unpurified
  • Cat. no.:
    89353-180

Specifications

About this item

This antibody reacts with B cell follicles in human lymphoid tissues. The mantle zones give a mosaic pattern, while the germinal centers show a coarse meshwork pattern of staining. This antibody stains free as well as bound kappa light chains. This antibody does not react with human lambda light chain.

Type: Primary
Antigen: Igkappa
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Customers Who Bought This Also Bought