Order Entry
ContactUsLinkComponent
Anti-ITK Goat Polyclonal Antibody
Anti-ITK Goat Polyclonal Antibody
Catalog #: 89362-840
Supplier:  Genetex
Anti-ITK Goat Polyclonal Antibody
Catalog #: 89362-840
Supplier:  Genetex
Price
$942.03
Log in for availability
Unit of Measure Each (50µG)
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    IL2-inducible T-cell kinase
  • Clonality:
    Polyclonal
  • Gene ID:
    3702
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    ITK
  • Conjugation:
    Unconjugated
  • Western blot:
    Yes
  • Antigen synonyms:
    EMT,
    kinase EMT,
    PSCTK2,
    interleukin-2-inducible T cell kinase,
    IL-2-inducible T cell kinase,
    T-cell-specific kinase,
    tyrosine-protein kinase ITK/TSK,
    homolog of mouse T-cell itk/tsk,
    LYK,
    tyrosine-protein kinase LYK
  • Storage buffer:
    0.5 mg/ml Tris saline containing 0.02% sodium azide and 0.5% BSA
  • Storage temperature:
    Upon receipt - Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    0.5 mg/ml
  • Immunogen:
    Peptide with sequence RLLRQLAEIAESGL, from C-Terminus of the protein sequence according to NP_005537
  • Tested applications:
    WB
  • Purification:
    Immunogen purified with antigen
  • Cat. no.:
    89362-840

Specifications

About this item

Goat polyclonal to ITK

For WB: Use at a concentration of 0.05-2 ?g/ml. A band at approximately 75 kDa is expected. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: ITK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human

Customers Who Bought This Also Bought