To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Immunoglobulin A
- Antigen symbol:IgA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Size:1 ml
- Immunogen:IgA isolated from a pool of normal human sera.
- Purification:purified immunoglobin fraction
- Storage buffer:PBS containing 0.1% sodium azide
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C.
- Tested applications:IHC-P, IHC-Fr
- Cat. no.:89351-804
- Supplier No.:GTX22411
Specifications
About this item
This antibody may be diluted to a titer of 1:25-1:100 in an ABC method. We suggest an incubation period of 30 minutes at room temperature. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining.
Type: Primary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human