Order Entry
ContactUsLinkComponent
Anti-GPCR GPR89 Rabbit Polyclonal Antibody
Anti-GPCR GPR89 Rabbit Polyclonal Antibody
Catalog #: 89366-462
Supplier:  Genetex
Anti-GPCR GPR89 Rabbit Polyclonal Antibody
Catalog #: 89366-462
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    G Protein-Coupled Receptor 88
  • Clonality:
    Polyclonal
  • Gene ID:
    54112
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    GPR88
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Size:
    25 μg
  • Antigen synonyms:
    STRG
  • Storage buffer:
    Phosphate buffered saline, pH 7.7, containing 0.01% sodium azide
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Synthetic peptide - KLH conjugated. Peptide sequence identity/similarity to Mouse: 84/89%.
  • Tested applications:
    IHC-P
  • Purification:
    Peptide immunogen affinity column
  • Cat. no.:
    89366-462
  • Supplier No.:
    GTX77979

Specifications

About this item

Rabbit polyclonal to G Protein-Coupled Receptor GPR89/SH120

Type: Primary
Antigen: GPR88
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Customers Who Bought This Also Bought