Order Entry
Canada
ContactUsLinkComponent
Anti-GPCR GPR105 Rabbit Polyclonal Antibody
Anti-GPCR GPR105 Rabbit Polyclonal Antibody
  89365-296
 :  Genetex
Anti-GPCR GPR105 Rabbit Polyclonal Antibody
  89365-296
 :  Genetex
 :  GTX71631

 

  • Antibody type:
    Primary
  • Antigen name:
    Purinergic Receptor P2Y, G-Protein Coupled, 14
  • Antigen symbol:
    P2Y14
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    25 μg
  • Gene ID:
    9934
  • Antigen synonyms:
    P2Y14|BPR105|GPR105
  • Storage buffer:
    Phosphate buffered saline, pH 7.7, containing 0.01% sodium azide
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Synthetic peptide - KLH conjugated
  • Tested applications:
    IHC-P
  • Purification:
    Affinity purified with antigen
  • Cat. no.:
    89365-296
  • Supplier No.:
    GTX71631

 

 

Rabbit Polyclonal to Human G Protein-Coupled Receptor GPR105

For IHC-P: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: P2RY14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human