To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Fibroblast Growth Factor 2 (Basic)
- Antigen symbol:FGF Basic
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:25μg
- Antigen synonyms:BFGF|HBGF-2|FGF-2|FGFB
- Gene ID:2247
- Immunogen:Recombinant human FGF basic
- Purification:Purified IgG prepared by affinity chromatography.
- Storage buffer:Phosphate buffered saline
- Storage temperature:Prior to reconstitution store at +4 °C. After reconstitution store at –20 °C. Storage in frost-free freezers is not recommended. This product should be storde undiluted. Should this product contain a precipitate we recommend microcentrifugation
- Tested applications:ELISA, WB
- Cat. no.:89359-462
- Supplier No.:GTX12476
Specifications
About this item
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members bind heparin and possess broad mitogenic and angiogenic activities. This protein has been implicated in diverse biological processes, such as limb and nervous system development, wound healing, and tumor growth. The mRNA for this gene contains multiple polyadenylation sites, and is alternatively translated from AUG and non-AUG (CUG) initiation codons resulting in five different isoforms with distinct properties. The CUG-initiated isoforms are localized in the nucleus and are responsible for the intracrine effect, whereas, the AUG-initiated form is mostly cytosolic and is responsible for the paracrine and autocrine effects of this FGF.
Recommended Starting Dilutions:ELISA: 0.15 - 0.30 ug/mlWestern Blotting: 0.1 - 0.2 ug/mlNot tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: FGF2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human