Order Entry
ContactUsLinkComponent
Anti-EIF2B Rabbit Polyclonal Antibody
Anti-EIF2B Rabbit Polyclonal Antibody
Catalog #: 89363-000
Supplier:  Genetex
Anti-EIF2B Rabbit Polyclonal Antibody
Catalog #: 89363-000
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Eukaryotic Initiation Factor 2 Beta
  • Clonality:
    Polyclonal
  • Gene ID:
    8893
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    EIF2B
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Size:
    100 μl
  • Western blot:
    Yes
  • Antigen synonyms:
    eukaryotic translation initiation factor 2 subunit beta,
    PPP1R67,
    EIF2,
    eukaryotic initiation factor 2-beta,
    EIF2beta,
    EIF2B,
    regulatory subunit 67,
    protein phosphatase 1,
    eukaryotic translation initiation factor 2 beta,
    eIF-2-beta,
    eukaryotic translation initiation factor 2 subunit 2
  • Storage buffer:
    PBS, pH7.4 containg 0.1% sodium azide
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    8.1 mg/ml
  • Immunogen:
    Synthetic peptide - Amino acids 50-65 of human eIF2B
  • Tested applications:
    ICC/IF, WB
  • Purification:
    IgG fraction
  • Cat. no.:
    89363-000
  • Supplier No.:
    GTX30808

Specifications

About this item

Rabbit polyclonal to eIF2B

For IF: Use at a dilution of 1:500-1:1,000. For WB: Use at a dilution of 1:500-1:1,000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: EIF2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Customers Who Bought This Also Bought