To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen symbol:EBNA 3C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:IgG
- Western blot:Yes
- Immunogen:Full length protein.
- Storage buffer:Preservative: 0.08% Sodium Azide. Constituents: PBS
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Tested applications:WB
- Cat. no.:89360-180
- Supplier No.:GTX16128
Specifications
About this item
EBNA 3C is a latent viral nuclear protein expressed in EBV transformed lymphoblastic cell lines and is essential for EBV mediated transformation of these cell lines. This viral nuclear protein is essential for EBV mediated positive immunoblastic lymphomas, and functions as a transcription factor.
Type: Primary
Antigen: EBNA 3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: