Order Entry
ContactUsLinkComponent
Anti-E6AP Rabbit Polyclonal Antibody
Anti-E6AP Rabbit Polyclonal Antibody
Catalog #: 89364-732
Supplier:  Genetex
RB PAB TO E6AP
Anti-E6AP Rabbit Polyclonal Antibody
Catalog #: 89364-732
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ubiquitin Protein Ligase E3A
  • Clonality:
    Polyclonal
  • Gene ID:
    7337
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    E6AP
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Size:
    50 μl
  • Western blot:
    Yes
  • Antigen synonyms:
    E6-AP,
    ANCR,
    EPVE6AP,
    AS,
    HPVE6A
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Immunogen:
    Recombinant protein encoding amino acids residues 1-851 of human E6-AP expressed in E. coli.
  • Tested applications:
    WB
  • Purification:
    Rabbit Polyclonal Antiserum
  • Cat. no.:
    89364-732
  • Supplier No.:
    GTX70169

Specifications

About this item

Rabbit Polyclonal antiserum to E6AP

Not tested in other applications. Optimal dilutions/concentations should be determined by the researcher.

Type: Primary
Antigen: E6AP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Customers Who Bought This Also Bought