Anti-MAPT Rabbit Polyclonal Antibody
Catalog # 89348-726
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Microtubule-Associated Protein tau
- Antigen symbol:MAPT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:4137
- Antigen synonyms:PPP1R103|FTDP-17|TAU|PPND|MAPTL|MTBT1|MTBT2|MSTD|DDPAC
- Storage buffer:1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular weight:79 kDa
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.75 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human Tau. The exact sequence is proprietary.
- Tested applications:WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89348-726
- Supplier No.:GTX100866
Specifications
About this item
Rabbit Polyclonal antibody to Tau (microtubule-associated protein tau)
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: MAPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human