Order Entry
ContactUsLinkComponent
Anti-CMA Mouse Monoclonal Antibody [clone: CC1]
Catalog #: 89367-130
Supplier:  Genetex
Price
$1,461.45
Log in for availability
Unit of Measure Each (1ml)
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Chymase 1
  • Clonality:
    Monoclonal
  • Clone:
    CC1
  • Gene ID:
    1215
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Antigen symbol:
    CMA
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Antigen synonyms:
    heart
    mast cell
    mast cell protease I
    chymase
    chymase 1 preproprotein transcript E
    chymase 1 preproprotein transcript I
    alpha-chymase
  • Storage buffer:
    Contains sodium azide as preservative
  • Storage temperature:
    Keep as concentrated solution, aliquot and store at 4°C.
  • Immunogen:
    BALB/C mice were injected with a purified human skin Chymase.
  • Tested applications:
    IHC-P
  • Purification:
    This antibody is supplied as purified immunoglobulin fraction containing sodium azide as preservative.
  • Cat. no.:
    89367-130
  • Size:
    1 ml

Specifications

About this item