Anti-HCV NS3 Mouse Monoclonal Antibody
Catalog # 89360-500
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen symbol:HCV NS3
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Storage buffer:Preservative: None. Constituents: 0.01M PBS. pH 7.2
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Purified recombinant chimeric HCV polyprotein (555 amino acids).
- Tested applications:ELISA, WB
- Purification:Protein G affinity purified
- Cat. no.:89360-500
- Supplier No.:GTX18668
Specifications
About this item
HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural proteins necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.
Type: Primary
Antigen: HCV NS3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: