Order Entry
ContactUsLinkComponent
Anti-FAK Rabbit Polyclonal Antibody
Anti-FAK Rabbit Polyclonal Antibody
Catalog #: 89361-514
Supplier:  Genetex
Anti-FAK Rabbit Polyclonal Antibody
Catalog #: 89361-514
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Tyrosine Kinase 2
  • Clonality:
    Polyclonal
  • Gene ID:
    5747
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    PTK2
  • Conjugation:
    Unconjugated
  • Size:
    50 μl
  • Western blot:
    Yes
  • Antigen synonyms:
    PTK2,
    FAK-related non-kinase polypeptide,
    protein-tyrosine kinase 2,
    FADK 1,
    focal adhesion kinase 1
  • Modification:
    phospho-Tyr577
  • Storage buffer:
    Preservative: 0.05% Sodium Azide; Constituents: PBS, 50% glycerol, 1mg/ml BSA (IgG, protease free). pH 7.4
  • Immunogen:
    Synthetic peptide derived from a region of Focal Adhesion Kinase that contains tyrosine 577. The sequence is conserved in human, mouse and frog.
  • Tested applications:
    WB
  • Purification:
    Immunogen affinity purified
  • Cat. no.:
    89361-514
  • Supplier No.:
    GTX24798

Specifications

About this item

Rabbit polyclonal to FAK ( phospho Y577 )

WB: Use at a concentration of 0.5 - 1.0 ?g/ml. Predicted molecular weight: 125 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PTK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Customers Who Bought This Also Bought