Order Entry
ContactUsLinkComponent
Anti-CSRP1 Rabbit Polyclonal Antibody
Anti-CSRP1 Rabbit Polyclonal Antibody
Catalog # 89358-002
Supplier:  Genetex
Anti-CSRP1 Rabbit Polyclonal Antibody
Catalog # 89358-002
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cysteine And Glycine-Rich Protein 1
  • Antigen symbol:
    CSRP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 μl
  • Antigen synonyms:
    CRP1|CRP|D1S181E|CSRP|CYRP|HEL-141
  • Gene ID:
    1465
  • Concentration:
    0.84 mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human CRP1. The exact sequence is proprietary.
  • Molecular weight:
    21 kDa
  • Purification:
    Purified by antigen-affinity chromatography.
  • Storage buffer:
    0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Storage temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Tested applications:
    WB
  • Cat. no.:
    89358-002
  • Supplier No.:
    GTX114344

Specifications

About this item

This gene encodes a member of the cysteine-rich protein (CSRP) family. This gene family includes a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. The LIM/double zinc-finger motif found in this gene product occurs in proteins with critical functions in gene regulation, cell growth, and somatic differentiation. [provided by RefSeq]

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CSRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Frequently Bought Together