Order Entry
ContactUsLinkComponent
Anti-CORO1A Goat Polyclonal Antibody
Anti-CORO1A Goat Polyclonal Antibody
  89360-066
 :  Genetex
Anti-CORO1A Goat Polyclonal Antibody
  89360-066
 :  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody type:
    Primary
  • Antigen name:
    Coronin 1A
  • Antigen symbol:
    CORO1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Bovine
  • Western blot:
    Yes
  • Size:
    50 μg
  • Antigen synonyms:
    HCORO1|IMD8|p57|CLABP|CLIPINA|TACO
  • Gene ID:
    11151
  • Concentration:
    0.5 mg/ml
  • Immunogen:
    Synthetic peptide: KRLDRLEETVQAK, corresponding to amino acids 449-461 of Human Coronin 1.
  • Purification:
    Immunogen affinity purified
  • Storage buffer:
    Preservative: 0.02% Sodium Azide; Constituents: 0.5% BSA, 0.5mg/ml Tris-saline. pH 7.3
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Tested applications:
    ELISA, WB
  • Supplier No.:
    GTX14787

 

 

Coronin 1 was first isolated from Dictyostelium, but similar proteins have been identified in many species and individual cell types. The coronin-like protein in yeast promotes actin polymerization and also interacts with microtubules. Dictyostelium mutants lacking coronin are impaired in cytokinesis and all actin-mediated processes. Analysis of coronin-GFP (green-fluorescent protein) fusions and knockout mutants shows that coronin participates in the remodelling of the cortical actin cytoskeleton that is responsible for phagocytosis and macropinocytosis. Likewise, in mammalian neutrophils, a coronin-like protein is also associated with the phagocytic apparatus. Coronin 1 binds actin (by similarity) and is found in noninfected macrophages, localised to the cortical microtubule network; in mycobacteria-infected macrophages, progressively relocalized and retained around the mycobacterial phagosomes. Coronin1/TACO is expressed in brain, thymus, spleen, bone marrow and lymph node. Low expression observed in lung and gut.

Peptide ELISA: Use at a dilution of 1/16000. WB: Use at a concentration of 0.25 - 1 ?g/ml. Detects a band of approximately 60 kDa in Molt-4 and Jurkat lysates (predicted molecular weight: 53 kDa according to NP_009005). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CORO1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Dog, Rat, Bovine

Frequently Bought Together