To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Rad21 Cohesin Complex Component
- Antigen symbol:Cohesin
- Clonality:Monoclonal
- Clone:CM110-2C10
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Antigen synonyms:SCC1|hHR21|CDLS4|MCD1|NXP1|HR21|HRAD21
- Gene ID:5885
- Concentration:1 mg/ml
- Immunogen:An N-terminal fusion protein of the C-terminal fragment of Scc1 subunit human cohesin (amino acids 544-631).
- Amino acid number:544 to 631
- Storage buffer:0.1 ml of protein G purified ascites (1 mg/ml) in PBS and 0.1% sodium azide.
- Storage temperature:Upon Receipt - Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Tested applications:WB
- Cat. no.:89362-942
- Supplier No.:GTX30641
Specifications
About this item
Mouse monclonal antibody to Cohesin (Scc1-subunit) (Rad21 Antibody)
Western Blot Suggested working dilutions: *Western Blot - 1:500 (ECL)Immunofluorescence - ND*The investigator should determine the optimal working dilution for a specific application.
Type: Primary
Antigen: Cohesin
Clonality: Monoclonal
Clone: CM110-2C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: