Order Entry
Canada
ContactUsLinkComponent
Anti-CGA Mouse Monoclonal Antibody [clone: LK2H10]
Anti-CGA Mouse Monoclonal Antibody [clone: LK2H10]
Catalog # 89353-232
Supplier:  Genetex
Anti-CGA Mouse Monoclonal Antibody [clone: LK2H10]
Catalog # 89353-232
Supplier:  Genetex
Supplier Number:  GTX73638

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Chromogranin A
  • Antigen symbol:
    CgA
  • Clonality:
    Monoclonal
  • Clone:
    LK2H10
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    6 ml
  • Gene ID:
    1113
  • Antigen synonyms:
    betagranin (N-terminal fragment of chromogranin A)|SP-I|chromogranin-A|pituitary secretory protein I|parathyroid secretory protein 1
  • Storage temperature:
    Keep as concentrated solution, aliquot and store at 4°C.
  • Immunogen:
    Purified C-terminal half of chromogranin A from urine of patients with carcinoid syndrome.
  • Tested applications:
    IHC-P
  • Cat. no.:
    89353-232
  • Supplier No.:
    GTX73638

Specifications

About this item

Monoclonal Mouse Antibody to Human Chromogranin A

This antibody has been pretitered and quality controlled to work on formalin-fixed paraffin-embedded as well as acetone fixed cryostat tissue sections. No further titration is required. We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation should be determined by the user. Suitable for formalin/paraffin tissues.

Type: Primary
Antigen: CGA
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: