Order Entry
ContactUsLinkComponent
Anti-CDK8 Rabbit Polyclonal Antibody
Catalog #: 89356-322
Supplier:  Genetex
Price
$920.76
Log in for availability
Unit of Measure Each (100µl)
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclin-dependent Kinase 8
  • Clonality:
    Polyclonal
  • Gene ID:
    1024
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
    Mouse
  • Antigen symbol:
    CDK8
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Antigen synonyms:
    Cell division protein kinase 8
    MGC126075
    MGC126074
    K35
    Protein kinase K35
  • Storage buffer:
    0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Molecular weight:
    53 kDa
  • Storage temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1mg/ml
  • Immunogen:
    Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human CDK8. The exact sequence is proprietary.
  • Tested applications:
    ELISA-Ag
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. no.:
    89356-322
  • Size:
    100 μl

Specifications

About this item