Order Entry
ContactUsLinkComponent
Anti-C5AR1 Mouse Monoclonal Antibody [clone: P12/1]
Anti-C5AR1 Mouse Monoclonal Antibody [clone: P12/1]
Catalog #: 89365-888
Supplier:  Genetex
Anti-C5AR1 Mouse Monoclonal Antibody [clone: P12/1]
Catalog #: 89365-888
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C5a Receptor
  • Clonality:
    Monoclonal
  • Clone:
    P12/1
  • Gene ID:
    728
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Antigen symbol:
    C5AR1
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Size:
    50 μg
  • Antigen synonyms:
    complement component 5a receptor 1|CD88|C5R1|C5A|C5AR|C5a receptor
  • Storage buffer:
    Phosphate buffered saline containing 0.09% Sodium Azide 1% Bovine Serum Albumin
  • Storage temperature:
    Store at +4 °C or at –20 °C if preferred. Store undiluted. Storage in frost free freezers is not recommended. Photosensitive. Should this product contain a precipitate we recommend microcentrifugation before use.
  • Immunogen:
    C5aR - peptide: Met1 - Asn31
  • Tested applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Purified IgG prepared by ion exchange chromatography.
  • Cat. no.:
    89365-888
  • Supplier No.:
    GTX75734

Specifications

About this item

Mouse monoclonal antibody [P12/1] to C5a R1

Recommended Starting Dilutions: Flow Cytometry: Max Dilution 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: C5AR1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Customers Who Bought This Also Bought