Order Entry
Canada
ContactUsLinkComponent
Anti-CD7 Mouse Monoclonal Antibody [clone: LT7]
Anti-CD7 Mouse Monoclonal Antibody [clone: LT7]
Catalog # 89365-544
Supplier:  Genetex
Anti-CD7 Mouse Monoclonal Antibody [clone: LT7]
Catalog # 89365-544
Supplier:  Genetex
Supplier Number:  GTX74732

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD7 molecule
  • Antigen symbol:
    CD7
  • Clonality:
    Monoclonal
  • Clone:
    LT7
  • Conjugation:
    Biotin
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Size:
    100 μg
  • Gene ID:
    924
  • Antigen synonyms:
    LEU-9|TP41|Tp40|GP40
  • Storage buffer:
    Phosphate buffered saline containing 0.09% Sodium Azide 1% Bovine Serum Albumin
  • Storage temperature:
    Store at +4 °C or at –20 °C if preferred. This product should be stored undiluted. Storage in frost free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
  • Immunogen:
    Normal human peripheral blood lymphocytes
  • Tested applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Purified IgG prepared by affinity chromatography on Protein A
  • Cat. no.:
    89365-544
  • Supplier No.:
    GTX74732

Specifications

About this item

Mouse monoclonal antibody [LT7] to CD7

Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: