Order Entry
ContactUsLinkComponent
Anti-CD44V4 Mouse Monoclonal Antibody [clone: VFF-11]
Anti-CD44V4 Mouse Monoclonal Antibody [clone: VFF-11]
Catalog #: 89365-732
Supplier:  Genetex
Anti-CD44V4 Mouse Monoclonal Antibody [clone: VFF-11]
Catalog #: 89365-732
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cd44 Molecule (Indian Blood Group)
  • Clonality:
    Monoclonal
  • Clone:
    VFF-11
  • Gene ID:
    960
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Antigen symbol:
    CD44V4
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Size:
    100 test
  • Antigen synonyms:
    MDU2|HUTCH-I|LHR|MC56|Pgp1|CDW44|MIC4|ECMR-III|HCELL|MDU3|IN|CSPG8
  • Storage buffer:
    Tris buffered saline containing 0.02% Sodium Azide 1% Bovine Serum Albumin
  • Storage temperature:
    Store at +4 °C or at –20 °C if preferred. Store undiluted. Storage in frost free freezers is not recommended. Photosensitive. Should this product contain a precipitate we recommend microcentrifugation before use.
  • Immunogen:
    GST fusion protein corresponding to the variable domains (v3 to v10) of human CD44.
  • Tested applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Purified IgG
  • Cat. no.:
    89365-732
  • Supplier No.:
    GTX75289

Specifications

About this item

Mouse monoclonal antibody [VFF-11] to CD44v4

Type: Primary
Antigen: CD44V4
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Customers Who Bought This Also Bought