Anti-CD4 Mouse Monoclonal Antibody [clone: Q4120]
Catalog # 89350-876
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:CD4 molecule
- Antigen symbol:CD4
- Clonality:Monoclonal
- Clone:Q4120
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:50 test
- Gene ID:920
- Antigen synonyms:L3T4|p55|CD4mut|Ly-4|W3/25
- Storage buffer:Preservative: 15mM Sodium Azide; Constituents: 1% BSA, 0.01M PBS. pH 7.4
- Storage temperature:Store at +4°C
- Concentration:0.1 mg/ml
- Immunogen:CD4-transfected mouse T-cell hybridoma, 3DT, followed by CD4+ human T-cell CEM cells.
- Tested applications:Fluorescence-Activated Cell Sorting
- Purification:Protein A purified
- Cat. no.:89350-876
- Supplier No.:GTX13437
Specifications
About this item
CD4 is a single chain transmembraneous glycoprotein which belongs to the immunoglobulin superfamily. The antigen is involved in binding to MHC class II molecules. The intracellular domain of the antigen is associated with p56lck protein tyrosine kinase.
- FACS: Use 10μl for 10^6 cells. FITC Conjugated Monoclonal Anti-Human CD4 may beused for:1. Identification, quantification and monitoring of helper/inducer T cells in peripheral blood, biological fluids, lymphoid organs, and other tissues.2. Analysis of T cell mediated cytotoxicity.3. Characterization of subtypes of T cell leukemias and lymphomas.4. Studies of T cells in health and disease. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: