Order Entry
ContactUsLinkComponent
Anti-CA1 Goat Polyclonal Antibody
Anti-CA1 Goat Polyclonal Antibody
Catalog # 89363-260
Supplier:  Genetex
Anti-CA1 Goat Polyclonal Antibody
Catalog # 89363-260
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Carbonic Anhydrase 1
  • Antigen symbol:
    CA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    1 mg
  • Antigen synonyms:
    HEL-S-11|Car1|CA-I|CAB
  • Gene ID:
    759
  • Concentration:
    10 mg/ml
  • Immunogen:
    Full length native protein (purified from Human Erythrocytes) (Human).
  • Purification:
    Protein G purified
  • Storage buffer:
    Preservative: 0.01% Sodium Azide. Constituents: 40% Glycerol, PBS. pH 7.2
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Tested applications:
    Conjugation, Dot Blot, Immunomicroscopy
  • Cat. no.:
    89363-260
  • Supplier No.:
    GTX40602

Specifications

About this item

Goat polyclonal to Carbonic Anhydrase I (Biotin)

Conjugation: Use at an assay dependent dilution. Dot: Use at an assay dependent dilution. ELISA: 1/2500 - 1/25000. IM: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human

Frequently Bought Together