To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Canine ParvoVirus
- Antigen symbol:Canine ParvoVirus
- Clonality:Monoclonal
- Clone:CPV1-2A1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Dog,Mink,Cat
- Purification:IgG fraction
- Storage buffer:PBS with 0.05% sodium azide
- Storage temperature:Store at +4 °C or at -20 °C if preferred. Storage in frost free freezers is not recommended.
- Tested applications:ELISA, IHC-P
- Cat. no.:89367-626
- Supplier No.:GTX27669
Specifications
About this item
Recognises canine parvovirus (CPV) and feline panleucopenia virus (FPV). CPV is reported to be a host range variant of FPV which emerged as a result of mutations of the viral capsid. Does not recognise canine adenovirus (type2); canine coronavirus; canine distemper; canine parinfluenza virus; feline leukaemia virus and feline immunodeficiency virus.
Type: Primary
Antigen: Canine ParvoVirus
Clonality: Monoclonal
Clone: CPV1-2A1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Cat, Dog, Mink