Order Entry
ContactUsLinkComponent
Anti-C4D Mouse Monoclonal Antibody [clone: 42654]
Anti-C4D Mouse Monoclonal Antibody [clone: 42654]
Catalog # 89367-734
Supplier:  Genetex
Anti-C4D Mouse Monoclonal Antibody [clone: 42654]
Catalog # 89367-734
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Complement Component 4D
  • Antigen symbol:
    C4D
  • Clonality:
    Monoclonal
  • Clone:
    10-11
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 μg
  • Antigen synonyms:
    CH|C4B1|C4B5|C4F|C4B12|C4B2|C4b|CPAMD3|C4B3|CO4
  • Gene ID:
    721
  • Immunogen:
    Native, from human plasma
  • Purification:
    Purified IgG prepared by affinity chromatography on Protein A
  • Storage buffer:
    Borate buffered saline containing 0.09% Sodium Azide (NaN3)
  • Storage temperature:
    Store at +4 °C or at –20 °C if preferred. Storage in frost-free freezers is not recommended. This product should be stored undiluted. Should this product contain a precipitate we recommend microcentrifugation before use.
  • Tested applications:
    ELISA, ICC/IF, IHC-P, IHC-Fr, WB
  • Cat. no.:
    89367-734
  • Supplier No.:
    GTX42598

Specifications

About this item

Mouse monoclonal antibody [10-11] to C4d

Recommended Starting Dilutions: ELISA: 1/5000 - 1/20000 Immunofluorescence: 1/250 - 1/600 Immunohistology - Frozen: 1/100 - 1/750 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: C4D
Clonality: Monoclonal
Clone: 42654
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Customers Who Bought This Also Bought