To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Adhesion G Protein-Coupled Receptor B2
- Antigen symbol:ADGRB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:25μg
- Antigen synonyms:BAI2|Brain-specific angiogenesis inhibitor 2
- Gene ID:576
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide conjugated to KLH.
- Purification:Immunogen affinity purified
- Storage buffer:Preservative: 0.01% Sodium Azide; Constituents: PBS. pH 7.7
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Tested applications:IHC-P
- Cat. no.:89359-630
- Supplier No.:GTX13306
Specifications
About this item
BAI1, a p53-target gene, encodes brain-specific angiogenesis inhibitor, a seven-span transmembrane protein and is thought to be a member of the secretin receptor family. Brain-specific angiogenesis proteins BAI2 and BAI3 are similar to BAI1 in structure, have similar tissue specificities and may also play a role in angiogenesis.
IHC-P: Use at a dilution of 1/100 - 1/200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: ADGRB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human