To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:B-cell activating factor
- Antigen symbol:BAFF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μg
- Antigen synonyms:CD257|ZTNF4|THANK|TALL-1|TNFSF20|DTL|TNFSF13B|TALL1|TNLG7A|BLYS
- Gene ID:10673
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide (Human). The peptide sequence is identical between human and mouse origin.
- Purification:Immunogen affinity purified
- Storage buffer:Preservative: 0.02% Sodium Azide; Constituents: PBS
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Tested applications:WB
- Cat. no.:89361-792
- Supplier No.:GTX25965
Specifications
About this item
Rabbit polyclonal to BAFF Receptor
WB: Use at a concentration of 2 - 5 ?g/ml. Predicted molecular weight: 19 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: BAFF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human