Order Entry
Canada
ContactUsLinkComponent
 
Anti-Bacterial Luciferase Rabbit Polyclonal Antibody
Catalog # 89360-840
Supplier:  Genetex
Anti-Bacterial Luciferase Rabbit Polyclonal Antibody
Catalog # 89360-840
Supplier:  Genetex
Supplier Number:  GTX20460

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Bacterial Luciferase
  • Antigen symbol:
    Bacterial Luciferase
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Bacteria
  • Western blot:
    Yes
  • Storage buffer:
    0.02M Potassium Phosphate, 0.15M NaCl (pH 7.2) with 1% BSA, 50% glycerol and 0.01% Sodium Azide
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Immunogen:
    Luciferase from Photobacterium fischerii
  • Tested applications:
    Immunomicroscopy
  • Purification:
    IgG fraction
  • Cat. no.:
    89360-840
  • Supplier No.:
    GTX20460

Specifications

About this item

Type: Primary
Antigen: Bacterial Luciferase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria