Order Entry
ContactUsLinkComponent
Anti-APRIL Mouse Monoclonal Antibody [clone: Aprily-1]
Anti-APRIL Mouse Monoclonal Antibody [clone: Aprily-1]
Catalog #: 89360-150
Supplier:  Genetex
Anti-APRIL Mouse Monoclonal Antibody [clone: Aprily-1]
Catalog #: 89360-150
Supplier:  Genetex
Price
$966.77
Log in for availability
Unit of Measure Each (50µG)
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    A Proliferation-inducing Ligand
  • Clonality:
    Monoclonal
  • Clone:
    Aprily-1
  • Gene ID:
    8741
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Antigen symbol:
    APRIL
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Size:
    50 μg
  • Western blot:
    Yes
  • Antigen synonyms:
    APRIL,
    TNFSF13,
    TALL2,
    TRDL-1,
    CD256,
    UNQ383/PRO715
  • Amino acid number:
    93 to 233
  • Storage buffer:
    Phosphate-buffered saline containing 0.02% sodium azide
  • Storage temperature:
    Upon Receipt - Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Recombinant human APRIL corresponding to amino acids 93-233, expressed in bacteria.
  • Tested applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Purified
  • Cat. no.:
    89360-150

Specifications

About this item

Members in the TNF superfamily regulate immune responses and induce apoptosis. A novel member in the TNF family was recently identified by several groups and designated APRIL (for A PRoliferation-Inducing Ligand), TALL2 (for TNF and ApoL-related Leukocyte-expressed Ligand 2), and TRDL 1a (for TNF related death ligand 1a) in human and mouse. Two receptors for APRIL were recently identified and designated TACI and BCMA. APRIL stimulates B and T cell proliferation, triggers humoral immune responses, activates NF-kB, and induces cell death. APRIL and its close relative of BlyS and their receptors BCMA and TACI are involved in diseases of autoimmunity and cancer.

For FACS: Use at a concentration of 10 ?g/ml. This antibody also works for intracellular FC staining. For WB: Use at a concentration of 1 ?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: APRIL
Clonality: Monoclonal
Clone: Aprily-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Customers Who Bought This Also Bought