Order Entry
Canada
ContactUsLinkComponent
Anti-TUBA1A Rabbit Polyclonal Antibody
Anti-TUBA1A Rabbit Polyclonal Antibody
Catalog # 89358-230
Supplier:  Genetex
Anti-TUBA1A Rabbit Polyclonal Antibody
Catalog # 89358-230
Supplier:  Genetex
Supplier Number:  GTX115048

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tubulin alpha 1a
  • Antigen symbol:
    TUBA1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    7846
  • Antigen synonyms:
    tubulin alpha-3 chain|B-ALPHA-1|tubulin alpha-4A chain|alpha 1|tubulin B-alpha-1|alpha|tubulin H2-alpha|tubulin|hum-a-tub1|tubulin alpha-1 chain|alpha-tubulin 3|alpha 3|brain-specific|TUBA1|H2-ALPHA|TUBA1/3/4|tubulin alpha-1A chain|TUBA3|hum-a-tub2|LIS3
  • Storage buffer:
    0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Molecular weight:
    50 kDa
  • Storage temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    0.24 mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human alpha Tubulin 1A. The exact sequence is proprietary.
  • Tested applications:
    ICC/IF, IHC-P, WB
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. no.:
    89358-230
  • Supplier No.:
    GTX115048

Specifications

About this item

Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The genes encoding these microtubule constituents belong to the tubulin superfamily, which is composed of six distinct families. Genes from the alpha, beta and gamma tubulin families are found in all eukaryotes. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species. This gene encodes alpha tubulin and is highly similar to mouse and rat Tuba1 gene. Northern blotting studies have shown that the gene expression is predominantly found in morphologically differentiated neurologic cells. This gene is one of three alpha-tubulin genes in a cluster on chromosome 12q. [provided by RefSeq]

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TUBA1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human