Order Entry
Canada
ContactUsLinkComponent
Anti-ACTN4 Rabbit Polyclonal Antibody
Anti-ACTN4 Rabbit Polyclonal Antibody
Catalog # 89349-162
Supplier:  Genetex
undefined
Anti-ACTN4 Rabbit Polyclonal Antibody
Catalog # 89349-162
Supplier:  Genetex
Supplier Number:  GTX101669

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Actinin alpha 4
  • Antigen symbol:
    ACTN4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    81
  • Antigen synonyms:
    FSGS|F-actin cross-linking protein|Alpha-actinin-4|ACTININ-4|Non-muscle alpha-actinin 4|FSGS1
  • Storage buffer:
    0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Molecular weight:
    105 kDa
  • Storage temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    0.32mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the C-terminus region of human alpha Actinin 4. The exact sequence is proprietary.
  • Tested applications:
    ICC/IF, IHC-P, WB
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. no.:
    89349-162
  • Supplier No.:
    GTX101669

Specifications

About this item

Rabbit Polyclonal antibody to alpha Actinin 4 (actinin, alpha 4)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ACTN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human