To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Thyroid Hormone Receptor Alpha
- Antigen symbol:THRA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:500 μl
- Antigen synonyms:CHNG6|ERBA1|AR7|NR1A1|THRA2|EAR7|c-ERBA-1|ERB-T-1|THRA1|ERBA
- Gene ID:7067
- Immunogen:Synthetic peptide: GDETQWPNGVKRKRKNSQCS, corresponding to amino acids 13-32 of Human Thyroid Hormone Receptor alpha 1.
- Purification:Immunogen affinity purified
- Storage buffer:Phosphate-buffered saline, pH 7.4 containing 1% BSA and 0.09% sodium azide
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Tested applications:IHC-P
- Cat. no.:89351-170
- Supplier No.:GTX15543
Specifications
About this item
Thyroid receptors have expression patterns that vary according to tissue and developmental stage. Almost all tissues express the alpha-1, alpha-2 and beta-1 isoforms, but beta-2 is synthesized almost exclusively in hypothalamus, anterior pituitary and developing ear. Receptor alpha-1 is the first isoform expressed in the conceptus.
IHC-P: Use at a dilution of 1/100. Staining of formalin fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0 for 10 min followed by cooling at room temperature for 20 min. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: THRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Pig, Rat